Product Details

SNP ID
rs13070107
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:132656450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCTATACTAATTCACACCCCT[A/T]TATGAGTCCCTGTTGCCCCATATCC
Phenotype
MIM: 614288 MIM: 610552
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
ACAD11 PubMed Links

Gene Details

Gene
ACAD11
Gene Name
acyl-CoA dehydrogenase family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032169.4 Intron NP_115545.3
Gene
NPHP3-ACAD11
Gene Name
NPHP3-ACAD11 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
UBA5
Gene Name
ubiquitin like modifier activating enzyme 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320210.1 Intron NP_001307139.1
NM_001321238.1 Intron NP_001308167.1
NM_001321239.1 Intron NP_001308168.1
NM_024818.4 Intron NP_079094.1
NM_198329.3 Intron NP_938143.1

View Full Product Details