Product Details
- SNP ID
-
hCV31766908
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:14087624 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGGTGAAGATTAAGTGTTTGTGT[A/T]TGTTAAACTCAGTAAGTTTTATTTT
- Phenotype
-
MIM: 609463
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MKL2
PubMed Links
Gene Details
- Gene
- MKL2
- Gene Name
- MKL1/myocardin like 2
There are no transcripts associated with this gene.
View Full Product Details