Product Details
- SNP ID
-
rs12135649
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
30
- Location
-
Chr.1:205386775 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCTTACCACATACCCTTCCAGGAA[C/T]AGGGAAGCAAAGGCTCTTGAGGAAG
- Phenotype
-
MIM: 610480
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LEMD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78165917] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LEMD1
- Gene Name
- LEM domain containing 1
- Gene
- LEMD1-AS1
- Gene Name
- LEMD1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details