Product Details

SNP ID
rs10917096
Assay Type
Functionally tested
NCBI dbSNP Submissions
41
Location
Chr.1:21982215 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAAAGGGAGACCCAGACAAAAG[A/G]ACTGGAGTGACACGCAGGCAGGTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CELA3B PubMed Links

Gene Details

Gene
CELA3B
Gene Name
chymotrypsin like elastase family member 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007352.3 Intron NP_031378.1

View Full Product Details