Product Details
- SNP ID
-
hCV31831065
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:110468978 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGACGGAGCGTAGCTCTTCTGCA[G/T]AGCCGCCCGCTGTGGCCGGGACCGA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FAM216A
PubMed Links
Gene Details
- Gene
- FAM216A
- Gene Name
- family with sequence similarity 216 member A
There are no transcripts associated with this gene.
- Gene
- GPN3
- Gene Name
- GPN-loop GTPase 3
There are no transcripts associated with this gene.
View Full Product Details