Product Details

SNP ID
rs12331480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:86826883 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTTCAGAAAACAAACAAGCACCT[A/C]TGTGCAAGCTGGACTGGAATGGGGA
Phenotype
MIM: 613366
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC10A6 PubMed Links

Gene Details

Gene
SLC10A6
Gene Name
solute carrier family 10 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_197965.2 Intron NP_932069.1

View Full Product Details