Product Details

SNP ID
rs11207561
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:59992928 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAACAGTGGTCCTTTCATTTATGT[A/G]AACCTCATTCTTTGAGTTGGCCAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf87 PubMed Links

Gene Details

Gene
C1orf87
Gene Name
chromosome 1 open reading frame 87
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152377.2 Intron NP_689590.1
XM_017000307.1 Intron XP_016855796.1
XM_017000308.1 Intron XP_016855797.1

View Full Product Details