Product Details

SNP ID
rs12111306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:69679872 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCAAAAGCCCCAGGCAAAGTGGC[A/G]CTTAATATTAATTTTCTTCCTTTTT
Phenotype
MIM: 612625
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMBRD1 PubMed Links

Gene Details

Gene
LMBRD1
Gene Name
LMBR1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018368.3 Intron NP_060838.3
XM_011535941.1 Intron XP_011534243.1

View Full Product Details