Product Details

SNP ID
rs12110338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:13618923 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATCGGTTTTCATAAAAAAATAAA[C/G]ATCAGTTGAACACTTTGTCTTTGCA
Phenotype
MIM: 611533 MIM: 603854 MIM: 604483
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NOL7 PubMed Links

Gene Details

Gene
NOL7
Gene Name
nucleolar protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317724.1 Intron NP_001304653.1
NM_016167.4 Intron NP_057251.2
Gene
RANBP9
Gene Name
RAN binding protein 9
There are no transcripts associated with this gene.

Gene
SIRT5
Gene Name
sirtuin 5
There are no transcripts associated with this gene.

View Full Product Details