Product Details

SNP ID
rs12074652
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:3021754 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAATGGCTCGGGGTTCTGCAAAG[C/T]GGGCCTGTCTGGGGAGTTTGGACCC
Phenotype
MIM: 608535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTRT2 PubMed Links

Gene Details

Gene
ACTRT2
Gene Name
actin related protein T2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080431.4 273 Missense Mutation GCG,GTG A23V NP_536356.3

View Full Product Details