Product Details

SNP ID
rs11186914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92302874 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCAGTCATGTAAATACACTATTA[A/T]GGCACCATATATAATACAACAGCAG
Phenotype
MIM: 610637
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MARCH5 PubMed Links

Gene Details

Gene
MARCH5
Gene Name
membrane associated ring-CH-type finger 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017824.4 Intron NP_060294.1
XM_011539886.2 Intron XP_011538188.1

View Full Product Details