Product Details

SNP ID
rs11144209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:75062158 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGGGATATAATTCAAAAAGAAGA[A/C]AACCAACAAAGAGTAGGTGTAGATA
Phenotype
MIM: 608704
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NMRK1 PubMed Links

Gene Details

Gene
NMRK1
Gene Name
nicotinamide riboside kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127603.1 Intron NP_001121075.1
NM_017881.2 Intron NP_060351.1
XM_006717162.1 Intron XP_006717225.1
XM_006717163.2 Intron XP_006717226.1
XM_011518817.1 Intron XP_011517119.1
XM_011518818.1 Intron XP_011517120.1
XM_011518819.2 Intron XP_011517121.1
XM_017014868.1 Intron XP_016870357.1
XM_017014869.1 Intron XP_016870358.1
XM_017014870.1 Intron XP_016870359.1
XM_017014871.1 Intron XP_016870360.1
XM_017014872.1 Intron XP_016870361.1
XM_017014873.1 Intron XP_016870362.1
XM_017014874.1 Intron XP_016870363.1

View Full Product Details