Product Details
- SNP ID
-
rs11144211
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:75062732 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAATTACTTCCTCCCTTTTGCTTT[A/C]TATTAAACATTTAAAAAGGCTGAGC
- Phenotype
-
MIM: 608704
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NMRK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114509239] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NMRK1
- Gene Name
- nicotinamide riboside kinase 1
View Full Product Details