Product Details

SNP ID
rs11225051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:101894873 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGATAATTTTAGGAATAAAAAAAA[A/T]CTTACCAGCATTTCCTGAATACCGT
Phenotype
MIM: 607666
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANGPTL5 PubMed Links

Gene Details

Gene
ANGPTL5
Gene Name
angiopoietin like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178127.4 Intron NP_835228.2
XM_011542735.2 Intron XP_011541037.1
XM_017017466.1 Intron XP_016872955.1

View Full Product Details