Product Details

SNP ID
rs11213821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111293926 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCTGTAGTCAAATTTGGAGGGAA[C/T]AAACTACCCCTTTTCCTTATACTGA
Phenotype
MIM: 615693 MIM: 615694
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf53 PubMed Links

Gene Details

Gene
C11orf53
Gene Name
chromosome 11 open reading frame 53
There are no transcripts associated with this gene.

Gene
COLCA1
Gene Name
colorectal cancer associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302644.1 4962 UTR 3 NP_001289573.1
NM_001302645.1 4962 UTR 3 NP_001289574.1
NM_001302646.1 4962 UTR 3 NP_001289575.1
NM_001302647.1 4962 UTR 3 NP_001289576.1
NM_001302648.1 4962 UTR 3 NP_001289577.1
NM_207429.3 4962 UTR 3 NP_997312.1
Gene
COLCA2
Gene Name
colorectal cancer associated 2
There are no transcripts associated with this gene.

View Full Product Details