Product Details

SNP ID
rs11085086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4649841 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGGCGGCCCATCCAGCAATTCCC[A/G]TGCCTGCCCTGACCTCAGAGGGCGG
Phenotype
MIM: 606746 MIM: 615869
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYDGF PubMed Links

Gene Details

Gene
MYDGF
Gene Name
myeloid derived growth factor
There are no transcripts associated with this gene.

Gene
TNFAIP8L1
Gene Name
TNF alpha induced protein 8 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167942.1 Intron NP_001161414.1
NM_152362.2 Intron NP_689575.2
XM_005259487.3 Intron XP_005259544.1
XM_011527680.2 Intron XP_011525982.1

View Full Product Details