Product Details
- SNP ID
-
rs7592816
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:65092874 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTATGACGACGGTTTCCCCCATTC[A/G]AGTGATAGTGAATGAATTCTCACAA
- Phenotype
-
MIM: 616889
MIM: 179508
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CEP68
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28587286] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEP68
- Gene Name
- centrosomal protein 68
There are no transcripts associated with this gene.
- Gene
- RAB1A
- Gene Name
- RAB1A, member RAS oncogene family
View Full Product Details