Product Details

SNP ID
rs13195900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:143610662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAAGATGAGCTGCTTGGGACTC[C/T]GGCCCTTTTGGTGGGTGTGTGTACA
Phenotype
MIM: 608724
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHACTR2 PubMed Links

Gene Details

Gene
PHACTR2
Gene Name
phosphatase and actin regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100164.1 Intron NP_001093634.1
NM_001100165.1 Intron NP_001093635.1
NM_001100166.1 Intron NP_001093636.1
NM_014721.2 Intron NP_055536.2

View Full Product Details