Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001122842.2 | 124 | Intron | NP_001116314.1 | ||
NM_001199619.1 | 124 | Intron | NP_001186548.1 | ||
NM_001199620.1 | 124 | Intron | NP_001186549.1 | ||
NM_001199621.1 | 124 | Intron | NP_001186550.1 | ||
NM_001199622.1 | 124 | Intron | NP_001186551.1 | ||
NM_181782.4 | 124 | Intron | NP_861447.3 | ||
XM_005266822.4 | 124 | Intron | XP_005266879.1 | ||
XM_006715340.3 | 124 | Intron | XP_006715403.1 | ||
XM_011535455.2 | 124 | UTR 5 | XP_011533757.1 | ||
XM_017010269.1 | 124 | Intron | XP_016865758.1 | ||
XM_017010270.1 | 124 | Intron | XP_016865759.1 | ||
XM_017010271.1 | 124 | UTR 5 | XP_016865760.1 | ||
XM_017010272.1 | 124 | Intron | XP_016865761.1 | ||
XM_017010273.1 | 124 | Intron | XP_016865762.1 | ||
XM_017010274.1 | 124 | Intron | XP_016865763.1 |