Product Details

SNP ID
rs4251435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:43762948 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGATTCAAAAGAGGTTTCTCTCT[A/C]TTCAAGGCTGATTTCTTCACCCTAT
Phenotype
MIM: 606883
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IRAK4 PubMed Links

Gene Details

Gene
IRAK4
Gene Name
interleukin 1 receptor associated kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114182.2 Intron NP_001107654.1
NM_001145256.1 Intron NP_001138728.1
NM_001145257.1 Intron NP_001138729.1
NM_001145258.1 Intron NP_001138730.1
NM_016123.3 Intron NP_057207.2
XM_005268943.3 Intron XP_005269000.1
XM_005268944.4 Intron XP_005269001.1
XM_005268945.4 Intron XP_005269002.1
XM_005268947.4 Intron XP_005269004.1
XM_005268948.2 Intron XP_005269005.1
XM_005268949.2 Intron XP_005269006.1
XM_006719438.3 Intron XP_006719501.1
XM_006719439.2 Intron XP_006719502.1
XM_006719440.2 Intron XP_006719503.1
XM_006719441.2 Intron XP_006719504.1
XM_006719442.2 Intron XP_006719505.1
XM_006719443.3 Intron XP_006719506.1
XM_011538431.2 Intron XP_011536733.1
XM_011538432.1 Intron XP_011536734.1
XM_011538433.2 Intron XP_011536735.1
XM_017019390.1 Intron XP_016874879.1
XM_017019391.1 Intron XP_016874880.1
Gene
PUS7L
Gene Name
pseudouridylate synthase 7 like
There are no transcripts associated with this gene.

View Full Product Details