Product Details
- SNP ID
-
rs4489914
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:34574709 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTTTCCAAGACATTTTACCTGAA[A/G]AAATATACCAAATTCATTTTTTTTT
- Phenotype
-
MIM: 606098
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNX6
PubMed Links
Gene Details
- Gene
- SNX6
- Gene Name
- sorting nexin 6
View Full Product Details