Product Details

SNP ID
rs4570511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:808892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACTACACAATAATTAAACACCG[C/T]AAAAGATACTATTCTAAATAAAGAG
Phenotype
MIM: 616513
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LARP4B PubMed Links

Gene Details

Gene
LARP4B
Gene Name
La ribonucleoprotein domain family member 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015155.2 6568 UTR 3 NP_055970.1

View Full Product Details