Product Details

SNP ID
rs3218538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:152648786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAAACATCCTGTGCTTCACCAG[C/T]TGCTGCCATGCCTTACAGAGATAAG
Phenotype
MIM: 600375
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
XRCC2 PubMed Links

Gene Details

Gene
XRCC2
Gene Name
X-ray repair cross complementing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005431.1 785 Silent Mutation CAA,CAG Q233Q NP_005422.1

View Full Product Details