Product Details

SNP ID
rs16960555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57826845 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTGGACCAACAGCGACCTGGCGG[C/T]GGTTCCACAGCTCTGGCTAGACCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FKBP1AP1 PubMed Links

Gene Details

Gene
FKBP1AP1
Gene Name
FK506 binding protein 1A pseudogene 1
There are no transcripts associated with this gene.

Gene
ZNF587B
Gene Name
zinc finger protein 587B
There are no transcripts associated with this gene.

View Full Product Details