Product Details

SNP ID
rs17854951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43797766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGATGGCTGTCCAGGGGCTGGTG[C/G]TGCCCTCGGTGGTGCAGGAGGGCCG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LYPD5 PubMed Links

Gene Details

Gene
LYPD5
Gene Name
LY6/PLAUR domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031749.2 712 Missense Mutation ACC,AGC T194S NP_001026919.2
NM_001288763.1 712 Missense Mutation ACC,AGC T151S NP_001275692.1
NM_182573.2 712 Missense Mutation ACC,AGC T151S NP_872379.2
XM_011526794.2 712 Missense Mutation ACC,AGC T151S XP_011525096.1

View Full Product Details