Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001081573.2 | Intron | NP_001075042.1 | |||
NM_001282283.1 | Intron | NP_001269212.1 | |||
NM_080612.3 | Intron | NP_542179.1 | |||
XM_005274648.1 | Intron | XP_005274705.1 | |||
XM_006724804.1 | Intron | XP_006724867.1 | |||
XM_011531103.1 | Intron | XP_011529405.1 | |||
XM_011531105.1 | Intron | XP_011529407.1 | |||
XM_011531106.1 | Intron | XP_011529408.1 | |||
XM_017029276.1 | Intron | XP_016884765.1 |