Product Details

SNP ID
rs16895878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17845658 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAATACTTCAAGATCAATTGATAA[A/C]TGCTTATTGTTCAGAAGGAATACTG
Phenotype
MIM: 611799 MIM: 606280
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LCORL PubMed Links

Gene Details

Gene
LCORL
Gene Name
ligand dependent nuclear receptor corepressor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166139.2 5875 Intron NP_001159611.1
NM_153686.8 5875 UTR 3 NP_710153.2
XM_011513821.2 5875 UTR 3 XP_011512123.1
XM_011513822.2 5875 UTR 3 XP_011512124.1
XM_011513823.2 5875 UTR 3 XP_011512125.1
XM_011513825.2 5875 Intron XP_011512127.1
XM_011513826.2 5875 UTR 3 XP_011512128.1
XM_017007961.1 5875 UTR 3 XP_016863450.1
XM_017007962.1 5875 UTR 3 XP_016863451.1
XM_017007963.1 5875 UTR 3 XP_016863452.1
XM_017007964.1 5875 Intron XP_016863453.1
XM_017007965.1 5875 UTR 3 XP_016863454.1
Gene
NCAPG
Gene Name
non-SMC condensin I complex subunit G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022346.4 5875 Intron NP_071741.2
XM_011513876.2 5875 Intron XP_011512178.1
XM_017008543.1 5875 Intron XP_016864032.1
XM_017008544.1 5875 Intron XP_016864033.1

View Full Product Details