Product Details

SNP ID
rs16941095
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:46717824 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGCGAACAAGGTGACGGAAATCA[C/T]GCTTAACAACTTTGACAAGGTCCTG
Phenotype
MIM: 601633
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NSF PubMed Links

Gene Details

Gene
NSF
Gene Name
N-ethylmaleimide sensitive factor, vesicle fusing ATPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006178.3 Intron NP_006169.2

View Full Product Details