Product Details

SNP ID
rs17852784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:46713914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTGCAGAGGAATCCAACTTCCC[A/G]TTCATCAAGATCTGTTCTCCTGATA
Phenotype
MIM: 601633
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NSF PubMed Links

Gene Details

Gene
NSF
Gene Name
N-ethylmaleimide sensitive factor, vesicle fusing ATPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006178.3 1796 Silent Mutation CCA,CCG P563P NP_006169.2

View Full Product Details