Product Details

SNP ID
rs16891774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:118955020 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACACTTCCTCACCTGAAAATTGGA[A/G]ATACCGGTTCTGGTTATGCAGAGAA
Phenotype
MIM: 607620 MIM: 602643
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COLEC10 PubMed Links

Gene Details

Gene
COLEC10
Gene Name
collectin subfamily member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324095.1 Intron NP_001311024.1
NM_006438.4 Intron NP_006429.2
XM_005250756.3 Intron XP_005250813.1
Gene
TNFRSF11B
Gene Name
TNF receptor superfamily member 11b
There are no transcripts associated with this gene.

View Full Product Details