Product Details

SNP ID
rs17630580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:97092072 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTGTGTTAGAAACAGGCTACCTG[A/G]GCTATGTCTGAGTTATGTTGGATCC
Phenotype
MIM: 610466
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIX1 PubMed Links

Gene Details

Gene
LIX1
Gene Name
limb and CNS expressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153234.4 3765 UTR 3 NP_694966.3

View Full Product Details