Product Details

SNP ID
rs17440396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:116751054 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAACTCAGCTCCCACATTTCATCC[A/G]TTAGGAAGCACTTCCTGATCACTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BUD13 PubMed Links

Gene Details

Gene
BUD13
Gene Name
BUD13 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159736.1 Intron NP_001153208.1
NM_032725.3 Intron NP_116114.1
XM_011543035.2 Intron XP_011541337.1

View Full Product Details