Product Details

SNP ID
rs17850650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103298678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTAGAAAGTGGACTCAGAGTAGC[C/T]TCGGAAGACTCTGGGCTCTCAACAT
Phenotype
MIM: 603131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PMPCB PubMed Links

Gene Details

Gene
PMPCB
Gene Name
peptidase, mitochondrial processing beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004279.2 244 Silent Mutation GCC,GCT A70A NP_004270.2
XM_005250717.3 244 Silent Mutation GCC,GCT A70A XP_005250774.1
XM_006716181.3 244 Silent Mutation GCC,GCT A70A XP_006716244.1

View Full Product Details