Product Details

SNP ID
rs17079684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:23982861 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAACCAGGTACACGTGTGAGCTTC[A/G]GGAGGCTGGGTCTCCCACCAGCCCT
Phenotype
MIM: 613324
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPATA13 PubMed Links

Gene Details

Gene
SPATA13
Gene Name
spermatogenesis associated 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166271.2 Intron NP_001159743.1
NM_001286792.1 Intron NP_001273721.1
NM_001286793.1 Intron NP_001273722.1
NM_001286794.1 Intron NP_001273723.1
NM_001286795.1 Intron NP_001273724.1
NM_153023.3 Intron NP_694568.1

View Full Product Details