Product Details

SNP ID
rs16975059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84975226 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGGGCGCCATCCTGGCCATTCCC[A/G]ACTGGAGCGCTGCCTGCGGTGGCAC
Phenotype
MIM: 614604
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZDHHC7 PubMed Links

Gene Details

Gene
ZDHHC7
Gene Name
zinc finger DHHC-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145548.1 1908 UTR 3 NP_001139020.1
NM_017740.2 1908 UTR 3 NP_060210.2
XM_011523222.1 1908 Intron XP_011521524.1
XM_017023434.1 1908 UTR 3 XP_016878923.1
XM_017023435.1 1908 UTR 3 XP_016878924.1
XM_017023436.1 1908 UTR 3 XP_016878925.1

View Full Product Details