Product Details
- SNP ID
-
rs17855377
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:50819969 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGTCTGTCACCTTCTGGACGTGG[A/G]CTTTTTTGCACTCATCATTAGGCAG
- Phenotype
-
MIM: 147910
MIM: 610601
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KLK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5517] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KLK1
- Gene Name
- kallikrein 1
- Gene
- KLK15
- Gene Name
- kallikrein related peptidase 15
There are no transcripts associated with this gene.
- Gene
- MGC45922
- Gene Name
- uncharacterized LOC284365
There are no transcripts associated with this gene.
View Full Product Details