Product Details
- SNP ID
-
rs17173608
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:150339575 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCATTGCTATAGTCCAGTGCCCTCC[G/T]TGAGCCGACAGAGGGAACTGGGGCC
- Phenotype
-
MIM: 601973
MIM: 615252
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LRRC61
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114887174] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LRRC61
- Gene Name
- leucine rich repeat containing 61
There are no transcripts associated with this gene.
- Gene
- RARRES2
- Gene Name
- retinoic acid receptor responder 2
- Gene
- ZBED6CL
- Gene Name
- ZBED6 C-terminal like
There are no transcripts associated with this gene.
View Full Product Details