Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001297673.1 | Intron | NP_001284602.1 | |||
NM_001297674.1 | Intron | NP_001284603.1 | |||
NM_015203.4 | Intron | NP_056018.2 | |||
XM_005245033.2 | Intron | XP_005245090.1 | |||
XM_006711240.2 | Intron | XP_006711303.1 | |||
XM_011509344.2 | Intron | XP_011507646.1 | |||
XM_017000763.1 | Intron | XP_016856252.1 | |||
XM_017000764.1 | Intron | XP_016856253.1 | |||
XM_017000765.1 | Intron | XP_016856254.1 | |||
XM_017000766.1 | Intron | XP_016856255.1 | |||
XM_017000767.1 | Intron | XP_016856256.1 |