Product Details
- SNP ID
-
rs16972021
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37153648 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGGTTACTATAGGAAGACATTTTA[G/T]CATTTGGTAATCTTTTTAACATTCT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF585A
PubMed Links
Gene Details
- Gene
- ZNF585A
- Gene Name
- zinc finger protein 585A
View Full Product Details