Product Details

SNP ID
rs17025292
Assay Type
Functionally Tested
NCBI dbSNP Submissions
37
Location
Chr.1:109988250 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAGAGTTCTAAAATAATTTTTCA[C/T]GGGAATGTCTTAATAGCTGTAGTGT
Phenotype
MIM: 607826
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AHCYL1 PubMed Links
Additional Information
For this assay, SNP(s) [rs145566124] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
AHCYL1
Gene Name
adenosylhomocysteinase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242673.1 Intron NP_001229602.1
NM_001242674.1 Intron NP_001229603.1
NM_001242675.1 Intron NP_001229604.1
NM_001242676.1 Intron NP_001229605.1
NM_006621.5 Intron NP_006612.2
XM_011540535.1 Intron XP_011538837.1

View Full Product Details