Product Details

SNP ID
rs17491190
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:202364101 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAGAAATATGAGGCCTAGAAAAA[A/G]GCTAAAGGAATACCTTAAGCATGGT
Phenotype
MIM: 603768
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R12B PubMed Links

Gene Details

Gene
PPP1R12B
Gene Name
protein phosphatase 1 regulatory subunit 12B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167857.1 Intron NP_001161329.1
NM_001167858.1 Intron NP_001161330.1
NM_001197131.1 Intron NP_001184060.1
NM_002481.3 Intron NP_002472.2
NM_032103.2 Intron NP_115286.1
NM_032104.2 Intron NP_115287.1
XM_005245206.4 Intron XP_005245263.1
XM_017001342.1 Intron XP_016856831.1
XM_017001343.1 Intron XP_016856832.1
XM_017001344.1 Intron XP_016856833.1
XM_017001345.1 Intron XP_016856834.1
XM_017001346.1 Intron XP_016856835.1
XM_017001347.1 Intron XP_016856836.1
XM_017001348.1 Intron XP_016856837.1
XM_017001349.1 Intron XP_016856838.1
XM_017001350.1 Intron XP_016856839.1
XM_017001351.1 Intron XP_016856840.1
XM_017001352.1 Intron XP_016856841.1
XM_017001353.1 Intron XP_016856842.1
XM_017001354.1 Intron XP_016856843.1
XM_017001355.1 Intron XP_016856844.1

View Full Product Details