Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_144579.2 | 3703 | UTR 3 | NP_653180.1 | ||
XM_005264648.1 | 3703 | UTR 3 | XP_005264705.1 | ||
XM_006712147.1 | 3703 | UTR 3 | XP_006712210.1 | ||
XM_011533186.2 | 3703 | UTR 3 | XP_011531488.1 | ||
XM_011533187.2 | 3703 | UTR 3 | XP_011531489.1 | ||
XM_011533188.2 | 3703 | UTR 3 | XP_011531490.1 | ||
XM_011533189.1 | 3703 | UTR 3 | XP_011531491.1 | ||
XM_011533190.2 | 3703 | UTR 3 | XP_011531492.1 | ||
XM_011533192.2 | 3703 | UTR 3 | XP_011531494.1 | ||
XM_011533194.2 | 3703 | UTR 3 | XP_011531496.1 | ||
XM_011533195.2 | 3703 | UTR 3 | XP_011531497.1 | ||
XM_011533196.2 | 3703 | UTR 3 | XP_011531498.1 | ||
XM_011533197.1 | 3703 | UTR 3 | XP_011531499.1 | ||
XM_017005340.1 | 3703 | UTR 3 | XP_016860829.1 | ||
XM_017005341.1 | 3703 | UTR 3 | XP_016860830.1 | ||
XM_017005342.1 | 3703 | UTR 3 | XP_016860831.1 | ||
XM_017005343.1 | 3703 | UTR 3 | XP_016860832.1 | ||
XM_017005344.1 | 3703 | UTR 3 | XP_016860833.1 | ||
XM_017005345.1 | 3703 | UTR 3 | XP_016860834.1 | ||
XM_017005346.1 | 3703 | UTR 3 | XP_016860835.1 |