Product Details

SNP ID
rs12118542
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:42384875 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCCAGACCTTGCACTAAAATTCC[A/C]TCTGCCTCAGATGCCCACTGTTCCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RIMKLA PubMed Links

Gene Details

Gene
RIMKLA
Gene Name
ribosomal modification protein rimK like family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173642.3 Intron NP_775913.2
XM_006710585.3 Intron XP_006710648.1
XM_017001082.1 Intron XP_016856571.1

View Full Product Details