Product Details

SNP ID
rs17070184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109115894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTGGTAATACTTTTCAAATGTT[A/G]AGTTGATATTTTTAGATTTTGCCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP57L1 PubMed Links

Gene Details

Gene
CEP57L1
Gene Name
centrosomal protein 57 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083535.1 Intron NP_001077004.1
NM_001271852.1 Intron NP_001258781.1
NM_001271853.1 Intron NP_001258782.1
NM_173830.4 Intron NP_776191.1
XM_005266935.4 Intron XP_005266992.3
XM_005266936.4 Intron XP_005266993.3
XM_005266937.4 Intron XP_005266994.3
XM_005266938.4 Intron XP_005266995.3
XM_006715462.3 Intron XP_006715525.1
XM_011535759.2 Intron XP_011534061.1
XM_011535762.2 Intron XP_011534064.1
XM_011535763.2 Intron XP_011534065.2
XM_017010764.1 Intron XP_016866253.1
XM_017010765.1 Intron XP_016866254.1
XM_017010767.1 Intron XP_016866256.1
XM_017010768.1 Intron XP_016866257.1
XM_017010769.1 Intron XP_016866258.1
XM_017010770.1 Intron XP_016866259.1
XM_017010771.1 Intron XP_016866260.1
XM_017010772.1 Intron XP_016866261.1

View Full Product Details