Product Details

SNP ID
rs17768065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:24539725 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCAGCAGCTTGCGGGCGTGTTCT[C/T]GCGGTTCCGGGCCTCAAGGCGACGG
Phenotype
MIM: 600938
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBBP6 PubMed Links

Gene Details

Gene
RBBP6
Gene Name
RB binding protein 6, ubiquitin ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006910.4 139 UTR 5 NP_008841.2
NM_018703.3 139 UTR 5 NP_061173.1
NM_032626.5 139 UTR 5 NP_116015.2

View Full Product Details