Product Details

SNP ID
rs16988828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30607574 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTATATGTTTGACTCCTCACCAG[A/G]CAAGATCTCCGTTAATTCAGTCATT
Phenotype
MIM: 605819 MIM: 613441
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PES1 PubMed Links

Gene Details

Gene
PES1
Gene Name
pescadillo ribosomal biogenesis factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243225.1 Intron NP_001230154.1
NM_001282327.1 Intron NP_001269256.1
NM_001282328.1 Intron NP_001269257.1
NM_014303.3 Intron NP_055118.1
XM_006724173.1 Intron XP_006724236.1
XM_017028678.1 Intron XP_016884167.1
Gene
TCN2
Gene Name
transcobalamin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000355.3 Intron NP_000346.2
NM_001184726.1 Intron NP_001171655.1

View Full Product Details