Product Details

SNP ID
rs17029870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:30727658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTCTGAAATATTAGATATCCCT[C/T]TGCTGGCAAAACATCAGAGTGGGTA
Phenotype
MIM: 615601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GADL1 PubMed Links

Gene Details

Gene
GADL1
Gene Name
glutamate decarboxylase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207359.2 2197 UTR 3 NP_997242.2
XM_017006297.1 2197 UTR 3 XP_016861786.1
XM_017006298.1 2197 UTR 3 XP_016861787.1

View Full Product Details