Product Details

SNP ID
rs17074834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42813956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGCAAATTCTAAGAAGTTCACA[C/T]TTTACCCAAACTTAATGTAATAACG
Phenotype
MIM: 602648
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACKR2 PubMed Links

Gene Details

Gene
ACKR2
Gene Name
atypical chemokine receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001296.4 Intron NP_001287.2
Gene
HIGD1A
Gene Name
HIG1 hypoxia inducible domain family member 1A
There are no transcripts associated with this gene.

View Full Product Details