Product Details

SNP ID
rs17868314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233639828 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTATATCAATTGGTATGGTCATTC[G/T]TTTACATTTCAGTATTCTAATGTGT
Phenotype
MIM: 606435 MIM: 606433
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UGT1A10 PubMed Links

Gene Details

Gene
UGT1A10
Gene Name
UDP glucuronosyltransferase family 1 member A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019075.2 Intron NP_061948.1
Gene
UGT1A8
Gene Name
UDP glucuronosyltransferase family 1 member A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019076.4 Intron NP_061949.3

View Full Product Details