Product Details

SNP ID
rs17848281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:3512687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAGGACTATCAGACCTGACAGC[A/C]GCTGGACATCGAGGTCCTCACTGGG
Phenotype
MIM: 104225
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LRPAP1 PubMed Links

Gene Details

Gene
LRPAP1
Gene Name
LDL receptor related protein associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002337.3 1446 UTR 3 NP_002328.1

View Full Product Details